Learn More
Purolite™ Chromalite™ HPLC Column for RP and IEX Chromatography
Prepacked stainless steel columns with robust Chromalite™ synthetic polymers for analytical reverse-phase (RP-HPLC) or ion exchange high pressure liquid chromatography (IEXHPLC).
Marca: Purolite™ LS06950-602
Dettagli aggiuntivi : Peso : 0.09000kg
Descrizione
Analytical Reverse Phase chromatography (RP-HPLC) and ion-exchange chromatography (IEX-HPLC) resins for separation of proteins, peptides and oligonucleotides, and ideal for biomolecule purification and polishing.
- Uniform particle sizes
- Exceptional batch-to-batch consistency
- Simple scale-up from laboratory analyses through to full-scale plant operation
- Highly hydrophobic
- High chemical stability
- More robust than silica materials
- Pressure stability up to 50 bar
- Efficient regeneration
- Pre-packed in stainless steel HPLC columns
- Compliant with FDA regulation 21 CFR 173.65
- Compliant with ResAP(2004) 3
- Halal, Kosher and TSE/BSE/GMO free
Specifica
4 to 6 μm | |
200 to 300 Å | |
5 years from date of manufacture; Storage (at 2°C to 20°C) | |
Chromalite™ 5AD2 | |
Macroporous Polystyrene/Divinylbenzene Adsorbent |
1 to 14 pH | |
>500 m2/g | |
4.6 mm x 250 mm | |
Stainless Steel |