Learn More
Chromalite™ M Methacrylic Hydrophilic Macroporous Resins, 100 to 300 μm, Purolite™
Chromalite™ M resins are hydrophilic, highly porous with high mechanical stability, based on a rigid methacrylic backbone. Optimal for protein purification and separation.
Marca:  Chromalite LS04432-192
Dettagli aggiuntivi : Peso : 0.08000kg
Codice prodotto. 17089483
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Descrizione
Descrizione
Hydrophilic, macroporous resins based on a methacrylic backbone.
  • Ion exchange chromatography
  • Typical porosity of 1000Å suitable for protein purification (up to 250 kDa)
  • Suitable for large-scale applications
  • Excellent pressure-flow properties,designed for high performance, stability and reliability
  • Purification of peptides obtained by fermentation
  • High mechanical stability
  • Uniform particle size
  • Stable to CIP (up to 1M NaOH)
  • Negligible swelling
  • Compliant with ResAP(2004) 3
  • Halal, Kosher and TSE/BSE/GMO free
Specifica
Specifica
Wet
100 to 300 μm
100 mL
Sulfopropyl / Na+
Sulfopropyl Methacrylate Macroporous Strong Cation Exchanger
1000 Å
Chromalite™ MS/C