missing translation for 'onlineSavingsMsg'
Learn More

Chromalite™ M Methacrylic Hydrophilic Macroporous Resins, 100 to 300 μm, Purolite™

Chromalite™ M resins are hydrophilic, highly porous with high mechanical stability, based on a rigid methacrylic backbone. Optimal for protein purification and separation.

Marca:  Chromalite LS04062-94

Dettagli aggiuntivi : Peso : 0.40000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 17059463

Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.


Descrizione

Descrizione

Hydrophilic, macroporous resins based on a methacrylic backbone.

  • Ion exchange chromatography
  • Typical porosity of 1000Å suitable for protein purification (up to 250 kDa)
  • Suitable for large-scale applications
  • Excellent pressure-flow properties,designed for high performance, stability and reliability
  • Purification of peptides obtained by fermentation
  • High mechanical stability
  • Uniform particle size
  • Stable to CIP (up to 1M NaOH)
  • Negligible swelling
  • Compliant with ResAP(2004) 3
  • Halal, Kosher and TSE/BSE/GMO free
Specifica

Specifica

Iminodiacetic Acid Methacrylate Affinity Resin
100 to 300 μm
500 mL
Chromalite™ MIDA/C
1 to 14 pH
White to Off-white Spherical Beads
1000 Å
Storage a 2°C to 20°C
Wet
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali