Learn More
CIAPIN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12638-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
CIAPIN1 is a cytokine-induced inhibitor of apoptosis with no relation to apoptosis regulatory molecules of the BCL2 or CASP families. Expression of CIAPIN1 is dependent on growth factor stimulation.Specifica
| CIAPIN1 | |
| Polyclonal | |
| Unconjugated | |
| CIAPIN1 | |
| 2810413N20Rik, Anamorsin, CIAPIN1, DRE2, PRO0915 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 109006, 307649, 57019 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5XID1, Q6FI81, Q8WTY4 | |
| CIAPIN1 | |
| CIAPIN1 Fusion Protein Ag3333 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |