missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ CLS-Y

Used for the isolation of genomic DNA from yeast, algae and fungi. MP Biomedicals CLS-Y is a cell lysis solution. It is a component included in the FastDNA and FastDNA SPIN Kits.

Marca:  MP Biomedicals™ 116540411

Dettagli aggiuntivi : CAS : 151-21-3 Peso : 0.11000kg

Codice prodotto. 15202830

  • 120.84€ / 110mL
Spedizione stimata: 14-06-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Applications: CLS-Y is a cell lysis solution used for the isolation of genomic DNA from yeast, algae and fungi
Product Overview:  CLS-Y is a component included in the FastDNA and FastDNA SPIN Kits and is a cell lysis solution used for the isolation of genomic DNA from yeast, algae and fungi
Product Description: CLS-Y is a cell lysis solution used for the isolation of genomic DNA from yeast, algae and fungi
Recommended Use: FastPrep™ Instrument 
Specifica

Specifica

Store at room temperature.
Algae, Bacteria (Gram +/-), Fungi, Spores, Yeast
110 mL
Yeast and Fungi lysis buffer
Cell Lysis Solution for Yeast and Fungi (CLS-Y)
Isolation of genomic DNA from yeast, algae and fungi
Up to 500 mg per prep
Liquid
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali