Learn More
Emma Safety Footwear Clyde (Ruffneck) Safety Shoes
Looks and feels like a classic sneaker. The highly supportive reinforced ruff neck heel together with the shock absorbing sole keeps ankles, shins and joints free from stress and injury.
Marca: Emma Safety Footwear 976546-40
Dettagli aggiuntivi : Peso : 1.62000kg
Descrizione
- S3-Safety shoe with sole plate for wet work
- Nubuck / Oil Nubuck construction
- GripForce™ Easy Twist Outsole
- 3M™ Reflective Striping
- ESD certified
- Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifica
EN ISO 20345:2011, EN 12568:2010 | |
Black | |
Hydro-Tec™ Sanitized Silver | |
40 |
Laces | |
Clyde (Ruffneck) | |
Nubuck, Oiled Nubuck | |
Steel |