missing translation for 'onlineSavingsMsg'
Learn More

COG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 10767-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16897363

  • 121.38€ / 20µL

Spedizione stimata: 28-09-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

The protein encoded by this gene is one of eight proteins which form a Golgi-localized complex required for normal Golgi morphology and function. It is thought that this protein is required for steps in the normal medial and trans Golgi-associated processing of glycoconjugates and plays a role in the organization of the Golgi-localized complex.
Specifica

Specifica

COG1
Polyclonal
Unconjugated
COG1
CDG2G, COG complex subunit 1, COG1, KIAA1381, LDLB
Rabbit
Antigen Affinity Chromatography
RUO
16834, 303652, 9382
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.55 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8WTW3, Q9Z160
Cog1
COG1 Fusion Protein Ag1203
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali