missing translation for 'onlineSavingsMsg'
Learn More

Collagenase, Type I from Clostridium histolyticum, Thermo Scientific Chemicals

Catalog Number 15855448
Click to view available options
Quantità:
100 mg
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Contains mixture of collagenase, clostripain, trypsin, caseinase and other lytic enzymes

  • Contains mixture of collagenase, clostripain, trypsin, caseinase and other lytic enzymes
TRUSTED_SUSTAINABILITY
Concentrazione ≥125 U/mg
Componenti Contains mixture of collagenase, clostripain, trypsin, caseinase and other lytic enzymes
Content And Storage Store desiccated at 4°C
Sorgente Clostridium histolyticum
Quantità 1 g
Product Type Collagenase
Forma Lyophilized

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'