Learn More
Fisherbrand™ Combs for MultiSUB Maxi Horizontal Gel Electrophoresis Unit, 2mm Thick
For use in preparatory electrophoresis and high-throughput screening.
Marca: Fisherbrand™
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Recommended to scale-up nucleic acid volumes for preparatory techniques
- Maximise sample volume
- Height adjustable to give user full control over well depth and sample-loading volume
- Rigid back which prevents heat-induced warping
- Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
- 20-sample and 40-sample combs are Multichannel pipettor compatible
Specifica
Blue | |
Comb | |
1 Each | |
34 μL |
MultiSUB Maxi Horizontal Gel Electrophoresis Unit | |
30 | |
2 mm |