Learn More
Lenz Laborglasintrumente Condensers acc. to Allihn, for Extractors, with Olives
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Marca: Lenz Laborglasintrumente 05226005
Dettagli aggiuntivi : Peso : 0.25000kg
Descrizione
- Allihn condensers for condensing vapours
- With NS cone
- Water connection: olives
- Made of DURAN™ tubing
Specifica
NS 34/35 | |
Duran Glass |