missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(Ii) Acetate Monohydrate, Honeywell™
Click to view available options
Quantità:
500 g
1 kg
50 kg
6 x 1 kg
Descrizione
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifica
Materiale o nome chimico | Copper(Ii) Acetate Monohydrate |
Quantità | 1 kg |
Formula lineare | Cu(CO2CH3)2 · H2O |
Numero UN | 1759 |
Beilstein | 3730548 |
IUPAC Name | copper;diacetate;hydrate |
Peso formulazione | 199.65g/mol |
Percent Purity | ≥99% (iodometric) |
Densità | Vapor Density: 6.8 (vs air) |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'