missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COPS2 Mouse anti-Human, Mouse, Rat, Clone: 2H10E6, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66435-1-IG-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predomitly cytosolic. NAD(+)-dependent isocitrate dehydrogenases catalyze the allosterically regulated rate-limiting step of the tricarboxylic acid cycle. Each isozyme is a heterotetramer that is composed of two alpha subunits, one beta subunit, and one gamma subunit. The protein encoded by this gene is the beta subunit of one isozyme of NAD(+)-dependent isocitrate dehydrogenase. Three alternatively spliced transcript variants encoding different isoforms have been described for this gene.Specifica
COPS2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P61201, P61202, P61203 | |
COPS2 | |
COPS2 Fusion Protein Ag1415 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG1 |
Western Blot, Immunohistochemistry (Paraffin) | |
2H10E6 | |
Unconjugated | |
COPS2 | |
ALIEN, Alien homolog, COPS2, CSN2, SGN2, Signalosome subunit 2, TR interacting protein 15, TRIP 15, TRIP15 | |
Mouse | |
Protein G | |
RUO | |
12848, 261736, 9318 | |
-20°C | |
Liquid |