Learn More
Corning™ Puntali filtranti per pipette Isotip™
Ideal for applications where avoiding cross-contamination is critical, such as DNA amplification and radioisotope handling
Marca: Corning™ 4823
87.49 EUR valido fino al 2024-04-30
Usa il codice promo "21637" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 1.17000kg
Descrizione
- IsoTip filtered pipet tips feature an inert, hydrophobic barrier that prevents aerosolized contaminants from coming in contact with pipettor shafts. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website.)
- Ideal for applications where avoiding cross contamination is critical, such as DNA amplification and radioisotope handling.
- Packaged sterile.
- RNase-/DNase-free.
- Nonpyrogenic.
Specifica
1 - 200 μl | |
Sì | |
Rack | |
Rack | |
Yes | |
Polipropilene |
Filtered | |
Incolore | |
Filtro | |
Universale | |
Isotip | |
10, 50 e 100 μl |