Learn More
Cole-Parmer™ CPS Products VP4S Pro-Set Single Stage Vacuum Pump
CPS Products VP4S Pro-Set Single Stage Vacuum Pump include both single and two stage pump configurations ranging in size from 2 to 12 CFM. All units incorporate heavy-duty dual voltage motors that can operate on either 115 VAC or 230 VAC (50 or 60 Hz) power at the flick of a switch.
Marca:  Cole-Parmer™ 07164-86
Dettagli aggiuntivi : Peso : 9.97000kg
Codice prodotto. 15442733
  • 308.00€ / Pezzo
Spedizione stimata: 10-07-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Operate on either 115 VAC or 230 VAC (50 or 60 Hz) power
  • Steel reinforced carrying handles are over molded for comfort and proprietary hard rubber bases absorb vibration for smoother, quieter operation.
  • All units incorporate integrated gas ballast valves, multi-threaded connection ports and are overload protected.
Specifica
Specifica
10 micron
Pro-Set Single Stage Vacuum Pump
50/60 Hz
110/220 V
VP4S Pro-Set
3 CFM
Vacuum System