Learn More
Abeba™ Crawler ALU 34510 Shoes
ESD dissipative shoe is Sanitized™ treated. In cludes TPU sole with extremly high slip resistance.
Marca: Abeba™ 34510/43
Dettagli aggiuntivi : Peso : 1.24000kg
Descrizione
- CE EN ISO 20345:2011 S1 SRC
- Microfiber with breathable textile inlays
- White/Gray
- Aluminum toe cap
- Foamed PU midsole
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3577)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
- Reflective material
- Discontinued model
Specifica
CE EN ISO 20345:2011 S1 SRC | |
Microfiber | |
Slip-resistant | |
34510 |
White, Gray | |
43 | |
Thermoplastic Polyurethane | |
Aluminum |