Learn More
Abeba™ Crawler ALU 34571 Shoes
ESD dissipative shoe is Sanitized™ treated. In cludes TPU sole with extremly high slip resistance.
Marca: Abeba™ 34571/44
Dettagli aggiuntivi : Peso : 1.26000kg
Descrizione
- CE EN ISO 20345:2011 S1 SRC
- Velours with breathable textile inlays
- Black/blue
- Aluminum toe cap
- Foamed PU midsole
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3577)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
- Reflective material
- Scratch-resistant coating in toe and heel area
Specifica
CE EN ISO 20345:2011 S1 SRC | |
Velour | |
Slip-resistant | |
34571 |
Black | |
44 | |
Polyurethane | |
Aluminum |