Learn More
Abeba™ Crawler ALU 34573 Shoes
ESD dissipative shoe is Sanitized™ treated. In cludes TPU sole with extremly high slip resistance.
Marca: Abeba™ 34573/46
Dettagli aggiuntivi : Peso : 1.24600kg
Descrizione
- CE EN ISO 20345:2011 S2 SRC
- Smooth leather white
- Aluminum toe cap
- Foamed PU midsole
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3577)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
- Reflective material
- Discontinued model
- Scratch-resistant coating in toe and heel area
Specifica
CE EN ISO 20345:2011 S2 SRC | |
Leather | |
Slip-resistant | |
34573 |
White | |
46 | |
Polyurethane | |
Aluminum |