Learn More
Abeba™ Crawler ALU 34803 Shoes
ESD dissipative shoe is Sanitized™ treated. In cludes TPU sole with extremly high slip resistance.
Marca: Abeba™ 34803/45
Dettagli aggiuntivi : Peso : 1.24100kg
Descrizione
- CE EN ISO 20345:2011 S3 SRC
- Microfiber black/yellow
- Aluminum toe cap
- Foamed PU midsole
- Metalfree penetration resistance
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3577)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
- Reflective material
- Upper one piece
Specifica
CE EN ISO 20345:2011 S3 SRC | |
Microfiber | |
Slip-resistant | |
34803 |
Black | |
45 | |
Polyurethane | |
Aluminum |