Learn More
Abeba™ Crawler ALU 34813 Shoes
ESD dissipative shoe is Sanitized™ treated. In cludes TPU sole with extremly high slip resistance.
Marca: Abeba™ 34813/43
Dettagli aggiuntivi : Peso : 1.24700kg
Descrizione
- CE EN ISO 20345:2011 S1P SRC
- Microfiber with breathable textile inlays
- Black/Yellow
- Aluminum toe cap
- Foamed PU midsole
- Metalfree penetration resistance
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3577)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
- Reflective material
- Discontinued model
Specifica
CE EN ISO 20345:2011 S1P SRC | |
Microfiber | |
Slip-resistant | |
34813 |
Black, Yellow | |
43 | |
Polyurethane | |
Aluminum |