missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRISPR-Cas9 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB10252-100
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
CRISPR-Cas9 Monoclonal specifically detects CRISPR-Cas9 in Multi-Species samples. It is validated for Western Blot, Simple Western.
Specifica
| CRISPR-Cas9 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| Cas9, CRISPR, CRISPR/Cas9, CRISPR-associated endonuclease Cas9/Csn1, CRISPR-associated protein 9 nuclease, CRISPR-Cas9/Csn1, csn1, SPy_1046, SPy1046, SpyCas9 | |
| E. coli-derived Cas9, Asp2-Asp1368, Accession # Q99ZW2 | |
| 100 μg | |
| Primary | |
| Detects Cas9 in direct ELISAs and Western blots. | |
| Multi-species | |
| Purified |
| Western Blot | |
| 1013816 | |
| Western Blot 2 μg/mL, Simple Western 20 μg/mL | |
| Q99ZW2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 901176 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto