Learn More
Sartorius™ Cubis™ Precision Balance
Designed for weighing with maximum accuracy, without complex workflows.
Marca: Sartorius™ MSE3203S-000-DE
Dettagli aggiuntivi : Peso : 10.00000kg
Descrizione
- Large, high-contrast LCD
- Simple menu guidance with short-text prompts
- Clear structured keys
Specifica
Precision Balance | |
1 mg | |
1 g | |
140 mm |
Large, High Contrast LCD | |
±5 mg | |
140 mm |