Learn More
R&D Systems™ Cultrex In Vitro Angiogenesis Assay (Tube Formation)
Provides a high-throughput screening platform to test the effects of multiple inhibitors and inducers on endothelial cell tube formation
Marca:  R&D Systems™ 3470-096-K
Dettagli aggiuntivi : Peso : 0.58200kg
Codice prodotto. 18452030
  • 465.43€ / 96 tests
Spedizione stimata: 01-07-2024
per visualizzare lo stock disponibile
Includes: BME Reduced Growth Factor, 5mL; Calcein AM, 50ug; Cell Staining Solution, 15mL; 10mM Sulforaphane, 15uL
Descrizione
Descrizione
Angiogenesis is the process by which new capillaries arise from pre-existing vasculature. Pathogenic angiogenesis is a common feature of rheumatoid arthritis, age-related macular degeneration, atherosclerosis and cancer.
Quantitatively assesses the ability of endothelial cells to form a tube network in a reconstituted protein matrix
Calcein AM fluorescence when cleaved to form calcein, providing data on cell viability and tube network formation in real time
Cultrex Redu
Specifica
Specifica
The product requires storage at -20 to -70° C and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.