Learn More
3M™ CUNO™ Metal Screen Filter Cartridge
3M™ CUNO™ Metal Screen Filter Cartridges are made from stainless steel wire cloth for controlled particle removal efficiency.
Marca: 3M™ 7100144432
Dettagli aggiuntivi : Peso : 0.25000kg
Descrizione
- Strong mesh construction withstands high pressure differentials across the cartridge for filtration of high viscosity fluids
- Elements can be cleaned chemically or ultrasonically and reused
- Ideal for heavy viscosity liquids requiring high differential pressures to achieve specified flow rates
Specifica
MSL10B6A1T0100 SP GSK A0006306 |