missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXADR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11777-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The protein encoded by this gene is a type I membrane receptor for group B coxsackieviruses and subgroup C adenoviruses. Pseudogenes of this gene are found on chromosomes 15, 18, and 21.Specifica
CXADR | |
Polyclonal | |
Unconjugated | |
CXADR | |
CAR, CVB3 binding protein, CXADR, HCAR, HCVADR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13052, 1525, 89843 | |
-20°C | |
Liquid |
Western Blot | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P78310, P97792, Q9R066 | |
CXADR | |
CXADR Fusion Protein Ag2369 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |