missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCR7 Mouse anti-Human, Mouse, Clone: 4C3D7, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60216-1-IG-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This antibody recognizes endogenous CXCR7, which has an experimentally determined molecular weight of 50 kDa.
This gene encodes a member of the G-protein coupled receptor family. Although this protein was earlier thought to be a receptor for vasoactive intestinal peptide (VIP), it is now considered to be an orphan receptor, in that its endogenous ligand has not been identified. The protein is also a coreceptor for human immunodeficiency viruses (HIV). Translocations involving this gene and HMGA2 on chromosome 12 have been observed in lipomas.Specifica
CXCR7 | |
Monoclonal | |
1.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P25106, P56485 | |
ACKR3 | |
CXCR7 Fusion Protein Ag14247 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
4C3D7 | |
Unconjugated | |
ACKR3 | |
Chemokine orphan receptor 1, CMKOR1, CXC R7, CXCR 7, CXCR7, G protein coupled receptor 159, GPR159, RDC 1, RDC1 | |
Mouse | |
Protein G | |
RUO | |
12778, 57007 | |
-20°C | |
Liquid |