Learn More
Lenz Laborglasinstrumente™ Imbuti contagocce cilindrici con tappo in vetro
For the safe, easy and accurate transfer of fluids in the laboratory. Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels, with Glass Plug are manufactured from durable DURAN™ borosilicate glass.
Marca: Lenz Laborglasinstrumente™ 04153711
Dettagli aggiuntivi : Peso : 0.14000kg
Descrizione
- With NS socket and cone
- With scale (approx. volume/mL)
- With inscription label
- Corrosion-free and resistant to thermal shock
Specifica
100 mL | |
14/23 NS | |
Yes |
Borosilicate Glass | |
Without Pressure Equalizing Tube | |
Dropping funnel |