Learn More
ACROBiosystems Cynomolgus Siglec-10 Protein, His Tag
Marca: ACROBiosystems SI0-C52H5-100ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The siglecs (sialic acid-binding Ig-like lectins) are a distinct subset of the Ig superfamily with adhesion-molecule-like structure. We describe here a novel member of the siglec protein family that shares a similar structure including five Ig-like domains, a transmembrane domain, and a cytoplasmic tail containing two ITIM-signaling motifs. Siglec-10 was identified through database mining of an asthmatic eosinophil EST library. The Siglec-10-VAP-1 interaction seems to mediate lymphocyte adhesion to endothelium and has the potential to modify the inflammatory microenvironment via the enzymatic end products.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 60.2 kDa. The protein migrates as 66-90 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.Specifica
60.2 kDa | |
Cynomolgus Siglec-10, His Tag (SI0-C52H5) is expressed from human 293 cells (HEK293). It contains AA Thr 17 - Ala 549 (Accession # A0A2K5WB x 8-1). | |
-20°C | |
Siglec-10 | |
Cynomolgus Monkey | |
25 mM MES, 150 mM NaCl, pH5.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
100 μg | |
Thr 17 - Ala 549 | |
SIGLEC10,MGC126774,PRO940,Siglec10,SLG2 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |