missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome c Mouse anti-Human, Mouse, Rat, Clone: 2D8D11, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66264-1-IG-20UL
Weitere Details : Peso : 0.00010kg
Beschreibung
The origin recognition complex (ORC) is a highly conserved six subunits protein complex essential for the initiation of the DNA replication in eukaryotic cells. Studies in yeast demonstrated that ORC binds specifically to origins of replication and serves as a platform for the assembly of additional initiation factors such as Cdc6 and Mcm proteins. The protein encoded by this gene is a subunit of the ORC complex. Studies of a similar gene in Drosophila suggested a possible role of this protein in neuronal proliferation and olfactory memory. Alternatively spliced transcript variants encoding distinct isoforms have been reported for this gene.Spezifikation
Cytochrome c | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P62897, P62898, P99999 | |
CYCS | |
Cytochrome c Fusion Protein Ag24349 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG2a |
Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
2D8D11 | |
Unconjugated | |
CYCS | |
CYC, CYCS, Cytochrome c, cytochrome c, somatic, HCS, THC4 | |
Mouse | |
Protein A | |
RUO | |
13063, 25309, 54205 | |
-20°C | |
Liquid |