missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytoglobin Mouse anti-Human, Mouse, Porcine, Rat, Clone: 4C7E11, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60228-1-IG-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Cytoglobin is a ubiquitously expressed hexacoordinate hemoglobin that may facilitate diffusion of oxygen through tissues, scavenge nitric oxide or other reactive oxygen species, or serve a protective function during oxidative stress.Specifica
Cytoglobin | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8WWM9, Q921A4, Q9CX80 | |
CYGB | |
Cytoglobin Fusion Protein Ag4161 | |
150 μL | |
Primary | |
Human, Mouse, Porcine, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
4C7E11 | |
Unconjugated | |
CYGB | |
CYGB, cytoglobin, HGB, Histoglobin | |
Mouse | |
Protein G | |
RUO | |
100518861, 114757, 114886, 170520 | |
-20°C | |
Liquid |