Learn More
Epredia™ Lab Vision™ Cytokeratin 8/18 Ab-1, Mouse Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cytokeratin 8/18 Ab-1, Mouse Monoclonal Antibody.
Marca: Epredia™ MS-743-S
Descrizione
Keratin 8 belongs to the type B (basic) subfamily of high molecular weight keratins and exists in combination with cytokeratin 18 (type A; acidic subfamily of low molecular weight keratins).Host Species: Mouse
Clone: 5D3
Isotype: IgG2a
Species Reactivity: Human. Others not known.
Immunogen: Cytokeratins purified from breast cancer MCF-7 cells
Molecular Weight: 52.5 kDa (Cytokeratin 8) and 45 kDa (Cytokeratin 18)
Positive Control: Skin
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifica
| Cytokeratin 8/18 Ab-1 | |
| Monoclonal | |
| Unconjugated | |
| Mouse | |
| 1 mL | |
| Cytoskeletal Protein | |
| Human | |
| Concentrated |
| Immunohistochemistry (Paraffin) | |
| 5D3 | |
| 1:50-1:100 for 30 min RT | |
| Cytokeratins purified from breast cancer MCF-7 cells | |
| IVD | |
| Primary | |
| Store vial at 4deg.C. When stored at 2 to 8deg.C, | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.