missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEDD Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10816-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
DEDD encodes a protein that contains a death effector domain (DED). DED is a protein-protein interaction domain shared by adaptors, regulators and executors of the programmed cell death pathway. Overexpression of this gene was shown to induce weak apoptosis. Upon stimulation, this protein was found to translocate from cytoplasm to nucleus and colocalize with UBTF, a basal factor required for RNA polymerase I transcription, in the nucleolus. At least three transcript variants encoding the same protein have been found for this gene.Specifica
| DEDD | |
| Polyclonal | |
| Unconjugated | |
| DEDD | |
| CASP8IP1, DEDD, DEDD1, DEDPro1, DEFT, FLDED 1, FLDED1, KE05 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9191 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O75618 | |
| DEDD | |
| DEDD Fusion Protein Ag1195 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto