missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantità:
10 g
100 g
500 g
Descrizione
Dimethyl methylphosphonate is used as a catalyst and a reagent in organic synthesis for the conversion of esters to ketophosphonates. It is an additive for the synthesis of unsaturated polyester resin which has flame retardant high phosphorous and UV-cured epoxy acrylate. It finds application as hydraulic fluids as well.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dimethyl methylphosphonate is used as a catalyst and a reagent in organic synthesis for the conversion of esters to ketophosphonates. It is an additive for the synthesis of unsaturated polyester resin which has flame retardant high phosphorous and UV-cured epoxy acrylate. It finds application as hydraulic fluids as well.
Solubility
Miscible with water, ethanol, ether, alcohol, benzene, acetone and carbon tetrachloride. Immiscible with heavy mineral oil.
Notes
Incompatible materials strong oxidizing agents and strong bases.
Dimethyl methylphosphonate is used as a catalyst and a reagent in organic synthesis for the conversion of esters to ketophosphonates. It is an additive for the synthesis of unsaturated polyester resin which has flame retardant high phosphorous and UV-cured epoxy acrylate. It finds application as hydraulic fluids as well.
Solubility
Miscible with water, ethanol, ether, alcohol, benzene, acetone and carbon tetrachloride. Immiscible with heavy mineral oil.
Notes
Incompatible materials strong oxidizing agents and strong bases.
Specifica
Materiale o nome chimico | Dimethyl methylphosphonate |
Densità | 1.16 |
Punti di ebollizione | 180°C to 181°C |
Punto d'infiammabilità | 68°C (154°F) |
Indice di rifrazione | 1.413 |
Quantità | 10 g |
Beilstein | 878263 |
Indice di Merck | 14,3391 |
Informazioni di solubilità | Miscible with water,ethanol,ether,alcohol,benzene,acetone and carbon tetrachloride. Immiscible with heavy mineral oil. |
Peso formulazione | 124.08 |
Vedi altri risultati |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'