Learn More
Lenz Laborglasintrumente Distilling Links Acc. to Claisen, with Vacuum Receiver Adapter & GL Thread
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Marca:  Lenz Laborglasintrumente 05057211
Dettagli aggiuntivi : Peso : 0.50000kg
Codice prodotto. 10413692
  • 160.38€ / Pezzo
Spedizione stimata: 13-08-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
  • Distilling links acc. to DIN 12594
  • With Liebig condenser and head to Claisen
  • Thread connections: GL14 (water) and GL18 (vacuum)
  • With two sockets NS 14/23
  • For distillation thermometers of 55 mm built-in length
  • Made of DURAN™ tubing
Specifica
Specifica
160 mm
NS 14/23