Learn More
DL-N-Benzoyl-2-methylserine, 98%, Thermo Scientific™
Catalog Number p-64307
Click to view available options
Quantità:
1 g
5 g
7508-82-9
C11H13NO4
223.23
MFCD01863306
FVWQRKJTTRAXJP-NSHDSACASA-N
s-2-benzamido-3-hydroxy-2-methylpropanoic acid, n-benzoyl-2-methylserine, 2s-2-benzamido-3-hydroxy-2-methylpropanoic acid, s-2-methyl-2-benzoylamino-3-hydroxypropanoic acid, 2s-3-hydroxy-2-methyl-2-phenylformamido propanoic acid
7167581
(2S)-2-benzamido-3-hydroxy-2-methylpropanoic acid
CC(CO)(C(=O)O)NC(=O)C1=CC=CC=C1
7508-82-9
C11H13NO4
223.23
MFCD01863306
FVWQRKJTTRAXJP-NSHDSACASA-N
s-2-benzamido-3-hydroxy-2-methylpropanoic acid, n-benzoyl-2-methylserine, 2s-2-benzamido-3-hydroxy-2-methylpropanoic acid, s-2-methyl-2-benzoylamino-3-hydroxypropanoic acid, 2s-3-hydroxy-2-methyl-2-phenylformamido propanoic acid
7167581
(2S)-2-benzamido-3-hydroxy-2-methylpropanoic acid
CC(CO)(C(=O)O)NC(=O)C1=CC=CC=C1
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico DL-N-Benzoyl-2-methylserine
Nota nome 98%
Punto di fusione 153.0°C to 159.0°C
Colore White
Spettro a infrarossi Authentic
Intervallo percentuale saggio 98%
Imballaggio Glass bottle
Quantità 1 g
Informazioni di solubilità Solubility in water: .. Other solubilities: soluble in ethanol,aq. naoh,aq. na2co3
Peso formulazione 223.23
Percent Purity 98%
Forma fisica Crystalline Powder