missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMWD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA564364
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: EPGTPFSIGR FATLTLQERR DRGAEKEHKR YHSLGNISRG GSGGS Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
DMWD (DM1 Locus, WD Repeat Containing) is a Protein Coding gene. Diseases associated with DMWD include Myotonic Dystrophy 1 and Corticosteroid Allergy. An important paralog of this gene is WDR20. [GeneCards]Specifica
DMWD | |
Polyclonal | |
Unconjugated | |
Dmwd | |
D19S593E; Dm9; DMRN9; DMR-N9; DMWD; dystrophia myotonica linked gene, N9; dystrophia myotonica WD repeat-containing protein; dystrophia myotonica, WD repeat containing; dystrophia myotonica-containing WD repeat motif; dystrophia myotonica-containing WD repeat motif protein; gene59; Protein 59; Protein DMR-N9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1762 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q09019 | |
Dmwd | |
Recombinant protein corresponding to Human DMWD | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only