missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18727-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This antibody is specific to DNA2.
DNA2 is a conserved helicase/nuclease involved in themaintece of mitochondrial and nuclear DNA stability.Specifica
DNA2 | |
Polyclonal | |
Unconjugated | |
DNA2 | |
DNA2, DNA2L, KIAA0083 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1763, 327762 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P51530, Q6ZQJ5 | |
Dna2 | |
Peptide | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |