missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC2/MPP11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11971-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
EIF4EBP1 is one member of a family of translation repressor proteins. It directly interacts with eukaryotic translation initiation factor 4E (eIF4E), which is a limiting component of the multisubunit complex that recruits 40S ribosomal subunits to the 5′ end of mRNAs. Interaction of this protein with eIF4E inhibits complex assembly and represses translation. This protein is phosphorylated in response to various signals including UV irradiation and insulin signaling, resulting in its dissociation from eIF4E and activation of mRNA translation.Specifica
DNAJC2/MPP11 | |
Polyclonal | |
Unconjugated | |
DNAJC2 | |
DNAJC2, M phase phosphoprotein 11, MPP11, ZRF1, ZUO1, Zuotin related factor 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116456, 22791, 27000 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P54103, Q7TQ20, Q99543 | |
DNAJC2 | |
DNAJC2/MPP11 Fusion Protein Ag2594 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |