Learn More
Emma Safety Footwear Donovan (Ruffneck) Safety Shoes
Looks and feels like a classic sneaker. Highly supportive reinforced ruff neck heel together with the shock absorbing sole keeps ankles, shins and joints free from stress and injury.
Marca: Emma Safety Footwear 974546-43
Dettagli aggiuntivi : Peso : 1.75500kg
Descrizione
- S3-Safety shoe with sole plate for wet work
- Nubuck / Oil Nubuck construction
- GripForce™ Easy Twist Outsole
- 3M™ Reflective Striping
- ESD certified
- Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifica
EN ISO 20345:2011, EN 12568:2010 | |
Brown | |
Hydro-Tec™ Sanitized Silver | |
43 |
Laces | |
Donovan (Ruffneck) | |
Nubuck, Oiled Nubuck | |
Steel |