Learn More
Huber Kaeltmaschinenbau™ Drain Tap
Designed for easy filling of the temperature control unit with thermofluid directly from the canister. Huber Kaeltmaschinenbau™ Drain Tap is ventilated to promote proper dispensing.
Marca: Huber Kaeltmaschinenbau™ 31735
Dettagli aggiuntivi : Peso : 0.10000kg
Descrizione
- Specifically designed for use with thermofluids.
- Ventilated for proper dispensing.
- Tap and valve assembly.
Specifica
Size: 23mm | |
1 Each | |
Drain Tap | |
Thermofluids |