Learn More
dsDNA Antibody (rDSD/8266), Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP3-23931-20ug
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
dsDNA Monoclonal antibody specifically detects dsDNA in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifica
| dsDNA | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Double stranded DNA | |
| Nuclei of Burkitt's cells | |
| 20 μg | |
| Primary | |
| Store at 4°C. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| rDSD/8266 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
Titolo del prodotto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido