missing translation for 'onlineSavingsMsg'
Learn More
Learn More
dsRNA Mouse anti-Broad Species Reactivity, Clone: K2, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP3-11398-10ML
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
dsRNA Monoclonal antibody specifically detects dsRNA in Broad Species Reactivity samples. It is validated for ELISA, Immunofluorescence
Specifica
| dsRNA | |
| Monoclonal | |
| Unconjugated | |
| Culture supernatant (RPMI, 5% fetal calf serum) | |
| Mouse | |
| Unpurified | |
| RUO | |
| Multi-species | |
| Purified |
| ELISA, Immunofluorescence | |
| K2 | |
| ELISA, Immunofluorescence | |
| Double-stranded RNA | |
| Female DBA/2 mice were injected intraperitonially with a mixture of 50 ug L-dsRNA and 75 ug methylated bovine serum albumin, emulsified in complete Freund's adjuvant. After several boosts spleen cells were fused with Sp2/0-Agl4 myeloma cells to generate the hybridoma clone. | |
| 10 mL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgM κ |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto