Learn More
Thermo Scientific™ dU Phosphoramidite, standard grade, 20-400 finish bottle
dU CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyuridine (dU) for use in oligonucleotide synthesis. The incorporation of a deoxyuridine base into a DNA sequence can be utilized to induce mutagenic effects.
Marca: Thermo Scientific™ 27-1738-08
Dettagli aggiuntivi : Peso : 0.04120kg
Descrizione
dU CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyuridine (dU) for use in oligonucleotide synthesis. The incorporation of a deoxyuridine base into a DNA sequence can be utilized to induce mutagenic effects. Enzymatically the uracil is removed, creating an abasic site at the dU position, and thus generating site-specific breaks in the oligo. This product is supplied in a 30 mL amber bottle with a 20-400 Finish neck style, which is compatible with most Expedite Systems of oligonucleotide synthesizers.
Features:
• Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyuridine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula: C39H47N4O8P
• Base: dU
• Formula weight: 730.8 g/mol
• CAS number: 109389-30-2
• HPLC Purity: ≥98.0%; conforms to reference
• 31P NMR Purity: ≥98.0%; conforms to reference
Specifica
Expedite Synthesizers | |
Store in freezer (at or below -20°C). | |
Room Temperature | |
0.25 g | |
Standard | |
Powder |
dU CEP | |
Deoxyuridine (dU) | |
98% | |
Phosphoramidite | |
DNA |