Learn More
DWK Life Sciences DURAN™ Funnel, with short stem
A funnel enables a clean transfer and filtration of liquids.
Marca: DWK Life Sciences 213516104
87.97 EUR valido fino al 2024-12-31
Usa il codice promo "23258" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.51000kg
Descrizione
Transferring liquids and solids is a common working method. In this activity the funnel is used, which due to its shape can absorb a relatively large amount of liquid or solid and transfer it through a constriction into a vessel with a small opening. Funnels are available in different sizes and designs, selected according to their application.
- With short stem
- Due to its shape, it is suitable for liquid-solid filtration using a filter paper
- Fired-on, highly durable white print
Specifica
200 mm | |
Yes | |
1 Ea. |
Boro 3.3 | |
Decanting and filtration of liquids; The spillage of substances is avoided; Filtering | |
Non-DIN/ISO size |