Learn More
Lenz Laborglasinstrumente™ Duran Glass ISO Form Separating Funnels
Created from borosilicate glass. Lenz Laborglasinstrumente™ Duran Glass ISO Form Separating Funnels are used in liquid-liquid extractions to separate the components of a mixture into immiscible solvent phases of different densities.
Marca: Lenz Laborglasinstrumente™ B0463151
Dettagli aggiuntivi : Peso : 0.40200kg
Descrizione
- According to ISO
- Material: DURAN™ Borosilicate Glass
- For use with liquid extractions
Specifica
1000 mL | |
Duran Glass, Borosilicate Glass |