Learn More
DWK Life Sciences DURAN™ Tube with Screw Thread, with DIN thread, and standard ground cone
The threaded tube with thread and standard ground joint allows the bonding of various glass components.
Marca:  DWK Life Sciences 248407207
182.40 EUR valido fino al 2024-12-31
Usa il codice promo "23258" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.11000kg
Codice prodotto. 13595370
  • 192.00€ / pezzi 10
Spedizione stimata: 21-06-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
This threaded tube has a screw thread and a standard ground joint. Components of an apparatus can be connected via both ends and thus connected to each other.
  • Tight closure due to standard ground joint
  • With DIN thread
Specifica
Specifica
10/Pk.
Tube with screw threads
GL 14, with Cone NS 19/26