missing translation for 'onlineSavingsMsg'
Learn More

DUSP10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 11689-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898933

  • 121.38€ / 20µL

Spedizione stimata: 06-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. GPSN2 catalyzes the final step, reducing trans-2, 3-enoyl-CoA to saturated acyl-CoA. Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. TER catalyzes the final step, reducing trans-2, 3-enoyl-CoA to saturated acyl-CoA.
Specifica

Specifica

DUSP10
Polyclonal
Unconjugated
DUSP10
DUSP10, MAP kinase phosphatase 5, MKP 5, MKP5
Rabbit
Antigen Affinity Chromatography
RUO
11221, 63953, 63995
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9ESS0, Q9Y6W6
Dusp10
DUSP10 Fusion Protein Ag2302
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali