Learn More
3M™ Filtri Dustmaster™
Particulate filter for use with the Dustmaster Powered Air Respirator helps protects against dust, mist and fume.
Marca: 3M™ 7000006601
Dettagli aggiuntivi : Peso : 0.15980kg
Descrizione
- Combines with the 6000 Series Gas Vapor cartridges to provide particulate and gas/vapor protection
- Note: 3M 501 filter retainers needed to combine the 5000 and 6000 series
Specifica
P2 | |
6000 and 7000 series facemasks | |
1 Ea. |
Dust Filter | |
Gasket |