Learn More
ABEBA™ Dynamic Burgundy Occupational Clogs Ob ESD
Burgundy dynamic occupational clog with non-foldable heel strap, replaceable active comfort insole, slip resistant TPU outsole
Marca: ABEBA™ 37327/40
Dettagli aggiuntivi : Peso : 0.79000kg
Descrizione
- Breathable, lightweight, comfortable occupational clogs
- Comply to EN ISO 20347: 2012, OB SRC
- ESD dissipative according to DIN EN 61340
- Slip resistant TPU outsole
- Good cushioning
- Energy absorption in heel area
- Water repellent burgundy knitted textile
- Non foldable heel strap, adjustable
- Removable active comfort insole
Specifica
Burgundy | |
40 | |
Unisex | |
Water Repellent |
Knitted Fabric | |
ESD dissipative to EN 61340 Climate class 1, Comply to EN ISO 20347: 2012 OB SRC | |
Clogs | |
OB SRC ESD |