Learn More
ABEBA™ Dynamic Flow Burgundy Occupational Slip On O1
Occupational slip-on shoes, in breathable burgundy knitted fabric, replaceable active MediFit insole, slip resistant TPU outsole
Marca:  ABEBA™ 7374/43
Dettagli aggiuntivi : Peso : 0.83000kg
Codice prodotto. 17111615
  • 66.08€ / 1 paio
Spedizione stimata:
per visualizzare lo stock disponibile
Descrizione
Descrizione
  • Breathable, lightweight, comfortable
  • Comply to EN ISO 20347: 2012, O1 SRC
  • Slip resistant TPU outsole
  • Good cushioning
  • Energy absorption in heel area
  • Burgundy knitted fabric
  • Double elastic band fitting system
  • Removable active Medifit insole
Specifica
Specifica
Burgundy
43
Unisex
Water Repellent
Knitted Fabric
Comply to EN ISO 20347: 2012 O1 SRC
Slip-on
O1 SRC