Learn More
ABEBA™ Dynamic Flow White Occupational Sneakers O1 ESD
White occupational sneakers with replaceable active MediFit insole, slip resistant TPU outsole
Marca: ABEBA™ 37370/39
Dettagli aggiuntivi : Peso : 0.77000kg
Descrizione
- Breathable, lightweight, comfortable occupational sneakers
- Comply to EN ISO 20347: 2012, O1 SRC
- ESD dissipative according EN 61340 class climatic 1
- Slip resistant TPU outsole
- Good cushioning
- Energy absorption in heel area
- White perforated microfiber
- Breathable lining with silver ions
- Removable active Medifit insole
Specifica
White | |
39 | |
Women's | |
No |
Perforated Microfiber | |
Comply to EN ISO 20347: 2012 O1 SRC, ESD class climatic 1 according EN 61340 | |
Sneakers | |
O1 SRC ESD |